Search
Search

Tnfsf13b| Gene Symbol: Tnfsf13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033622.1 | NP_296371.1 | 4, 5 | 777 | ||
| GenBank mRNA | AF119383.1 | - | 4, 5 | 777 | ||
| AF352245.1 | - | 4, 5 | 781 | |||
| AK034121.1 | - | 4, 5 | 409 | |||
| AK155288.1 | - | 4, 5 | 556 | |||
| BC106841.1 | - | 4, 5 | 750 | 40045720 | ||