Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Trim16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053169.2 | NP_444399.2 | 1 | 452 | ||
GenBank mRNA | AF449496.1 | - | 1 | 203 | ||
AK053139.1 | - | 1 | 266 |