Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Def8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253783.1 | NP_001240712.1 | 14 | 2054 | ||
NM_001253784.1 | NP_001240713.1 | 13 | 2032 | |||
NM_001281803.1 | NP_001268732.1 | 12 | 2036 | |||
NM_054046.5 | NP_473387.1 | 12 | 1945 | |||
GenBank mRNA | AK028726.1 | - | 13 | 2003 | ||
AK046271.1 | - | 12 | 1906 | |||
AK050405.1 | - | 13 | 2003 | |||
AK081295.1 | - | 12 | 1935 | |||
AK087778.1 | - | 12 | 1917 | |||
AK129019.1 | - | 12 | 1987 | |||
AK133979.1 | - | 12 | 1920 | |||
AK150334.1 | - | 12 | 1881 | |||
AK150994.1 | - | 12 | 1881 | |||
AK180490.1 | - | 1 | 394 | |||
AK192566.1 | - | 1 | 334 | |||
BC003306.1 | - | 14 | 2005 | 3154712 |