Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc34a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080854.3 | NP_543130.2 | 6 | 561 | ||
GenBank mRNA | AB054999.1 | - | 6 | 541 | ||
BC125326.1 | - | 5 | 494 | 40129841 | ||
BC131997.1 | - | 5 | 494 | 40130274 |