Search
Search

Cacng6| Gene Symbol: Cacng6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133183.1 | NP_573446.1 | 1 | 263 | ||
| GenBank mRNA | AF361348.1 | - | 1 | 263 | ||
| BC115456.1 | - | 1 | 263 | 40100157 | ||
| BC115457.1 | - | 1 | 263 | 40100159 | ||