Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryzl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026994.1 | NP_081270.1 | 2, 3 | 325 | ||
NM_133679.2 | NP_598440.1 | 2, 3 | 325 | |||
GenBank mRNA | AK008846.1 | - | 2, 3 | 101 | ||
AK010433.1 | - | 2, 3 | 129 | |||
AK017995.1 | - | 2, 3 | 326 | |||
AK145827.1 | - | 2, 3 | 149 | |||
BC010479.1 | - | 2, 3 | 122 | 3968727 | ||
BC019387.1 | - | 1, 2 | 95 | 4164847 | ||
BY717916.1 | - | 2, 3 | 326 |