Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133704.4 | NP_598465.2 | 2 | 205 | ||
GenBank mRNA | AK007341.1 | - | 2 | 145 | ||
AK037540.1 | - | 2 | 140 | |||
AK044615.1 | - | 2 | 140 | |||
AK082516.1 | - | 2 | 205 | |||
BC057176.1 | - | 1 | 118 | |||
BC138969.1 | - | 1 | 119 | 8861991 | ||
BC138971.1 | - | 1 | 119 | 8861993 | ||
BC145477.1 | - | 1 | 121 | |||
BC145478.1 | - | 1 | 121 | 9054040 |