Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pycr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133705.2 | NP_598466.1 | 2, 3 | 338 | ||
GenBank mRNA | AK007535.1 | - | 2, 3 | 320 | ||
AK150756.1 | - | 2 | 1349 | |||
AK152302.1 | - | 2 | 1349 | |||
AK165618.1 | - | 2, 3 | 313 | |||
AK167622.1 | - | 2, 3 | 294 | |||
AK178957.1 | - | 45 | ||||
BC006882.1 | - | 2, 3 | 314 | 2647612 | ||
CJ154235.1 | - | 2, 3 | 337 |