Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mat2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199274.1 | NP_001186203.1 | 5 | 955 | ||
NM_134017.2 | NP_598778.1 | 5 | 759 | |||
GenBank mRNA | AB070267.1 | - | 5 | 726 | ||
AK075600.1 | - | 5 | 660 | |||
AK075947.1 | - | 5 | 694 | |||
AK153126.1 | - | 5 | 715 | |||
AK162087.1 | - | 5 | 694 | |||
BC003457.1 | - | 5 | 690 | 2648452 |