Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146025.1 | NP_001139497.1 | 11 | 2453 | ||
NM_001146026.1 | NP_001139498.1 | 11 | 2378 | |||
NM_001146027.1 | NP_001139499.1 | 11 | 2244 | |||
NM_134064.2 | NP_598825.2 | 11 | 2386 | |||
GenBank mRNA | AK147349.1 | - | 11 | 2379 | ||
AK160090.1 | - | 11 | 2241 | |||
AK160115.1 | - | 11 | 2241 | |||
AK161651.1 | - | 11 | 2370 | |||
AK171943.1 | - | 11 | 2239 | |||
AK172257.1 | - | 11 | 2240 | |||
BC017630.1 | - | 11 | 2179 | 4507289 | ||
BC035548.1 | - | 11 | 2201 | 4221284 |