siRNA Details
Gene Symbol: Gstt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133994.3 | NP_598755.1 | 5 | 1399 | ||
GenBank mRNA | AF508157.1 | - | 5 | 1399 | ||
AK138669.1 | - | 5 | 1056 | |||
AK154161.1 | - | 5 | 1105 | |||
AK168226.1 | - | 5 | 1048 | |||
BC003903.1 | - | 5 | 1027 | 3601446 | ||
BC057964.1 | - | 5 | 1354 | 5352325 | ||
BC062201.1 | - | 5 | 1356 | 4913425 |

More Information
Other Species Targeted

Panther Classification:
