Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161355.1 | NP_001154827.1 | 3 | 573 | ||
NM_001161356.1 | NP_001154828.1 | 3 | 346 | |||
NM_134249.5 | NP_599010.4 | 3 | 455 | |||
GenBank mRNA | AB009015.1 | - | 3 | 346 | ||
AF399827.1 | - | 3 | 307 | |||
AF399828.1 | - | 3 | 307 | |||
AK145630.1 | - | 3 | 334 | |||
AK163225.1 | - | 3 | 334 | |||
AK166877.1 | - | 3 | 327 | |||
BC025096.1 | - | 3 | 385 | 4158605 | ||
BC028829.1 | - | 3 | 417 | 5102129 | ||
BY731803.1 | - | 3 | 455 | |||
CB953226.1 | - | 3 | 576 |