Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ybx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011733.2 | NP_035863.1 | 4, 5 | 621 | ||
NM_139117.2 | NP_620817.2 | 4, 5 | 621 | |||
GenBank mRNA | AF246224.1 | - | 4, 5 | 463 | ||
AF248546.1 | - | 4, 5 | 443 | |||
AF248547.1 | - | 4, 5 | 443 | |||
AK003145.1 | - | 4, 5 | 555 | |||
AK076041.1 | - | 4, 5 | 560 | |||
BC027785.1 | - | 4, 5 | 632 | 5345031 | ||
BC048242.1 | - | 4, 5 | 609 | 6493766 | ||
BC062377.1 | - | 4, 5 | 609 | 6530404 | ||
D14485.1 | - | 4, 5 | 609 |