Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab40c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139154.2 | NP_631893.1 | 7 | 1043 | ||
GenBank mRNA | AF422144.1 | - | 6 | 996 | ||
AK133766.1 | - | 6 | 819 | |||
BC027008.1 | - | 7 | 1031 | 5364768 | ||
BC037461.1 | - | 8 | 1475 |