Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vash2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144879.2 | NP_659128.2 | 3 | 723 | ||
GenBank mRNA | AK045261.1 | - | 3 | 519 | ||
AK078596.1 | - | 3 | 592 | |||
AK140989.1 | - | 3 | 717 | |||
AK163193.1 | - | 3 | 499 | |||
BC024141.1 | - | 3 | 493 | 5149750 |