Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271443.1 | NP_001258372.1 | 7 | 634 | ||
NM_001271444.1 | NP_001258373.1 | 7 | 874 | |||
NM_144870.5 | NP_659119.2 | 7 | 877 | |||
GenBank mRNA | AK135069.1 | - | 7 | 635 | ||
AK149674.1 | - | 7 | 835 | |||
AK153182.1 | - | 7 | 821 | |||
AK153900.1 | - | 7 | 611 | |||
AK164901.1 | - | 7 | 611 | |||
AK167907.1 | - | 7 | 612 | |||
BC021616.1 | - | 7 | 595 | 5132866 | ||
BC086766.1 | - | 7 | 588 | 6820786 |