siRNA Details
Gene Symbol: Slmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144867.2 | NP_659116.1 | 7 | 701 | ||
GenBank mRNA | AK143220.1 | - | 7 | 685 | ||
BC019561.1 | - | 7 | 646 | 4459623 |

Gene Symbol: Slmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144867.2 | NP_659116.1 | 7 | 701 | ||
GenBank mRNA | AK143220.1 | - | 7 | 685 | ||
BC019561.1 | - | 7 | 646 | 4459623 |