Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ALDOB SIRNA Assays ›
          Gene Symbol:
          Aldob
          Gene Name:
          aldolase B, fructose-bisphosphate
          Gene Aliases:
          Aldo-2, Aldo2, BC016435
          Chromosome Location:
          Chr.4: 49535995 - 49549483 on Build GRCm38
          UniGene ID:
          Mm.482116
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 88992

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Aldob
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_144903.2 NP_659152.1 3 205
          GenBank mRNA AK135385.1 - 3 234
          AK149395.1 - 3 222
          AK149598.1 - 3 219
          AK167334.1 - 3 222
          AK167566.1 - 3 219
          AK168600.1 - 3 219
          AK168997.1 - 3 269
          BC016435.1 - 3 201 4241025
          BC022113.1 - 3 178 5098524
          BC024056.1 - 3 211 5100067
          BC024112.1 - 3 210 5102118
          BC026577.1 - 3 170 4989521
          BC030724.1 - 3 207 5100784
          BC030725.1 - 3 211 5101686
          BC034169.1 - 3 211 4975382
          BC034171.1 - 3 188 4972521
          BC034172.1 - 3 175 4974489
          BC034173.1 - 3 187 4974553
          BC036130.1 - 3 188 4974071
          BC036131.1 - 3 205 4974877
          BC036132.1 - 3 205 4975336
          BC036133.1 - 3 217 4975573

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          aldolase

          Gene Ontology Categories:

          Function(s) Process(es)

          fructose metabolic process
          glycolytic process
          NADH oxidation
          response to zinc ion
          fructose 1,6-bisphosphate metabolic process
          positive regulation of ATPase activity
          response to peptide hormone
          vacuolar proton-transporting V-type ATPase complex assembly
          catalytic activity
          fructose-bisphosphate aldolase activity
          cytoskeletal protein binding
          lyase activity
          phosphatidylcholine binding
          identical protein binding
          ATPase binding
          fructose-1-phosphate aldolase activity
          fructose binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0