Search
Search

Map7d1| Gene Symbol: Map7d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145970.1 | NP_001139442.1 | 3 | 673 | ||
| NM_144941.3 | NP_659190.3 | 3 | 673 | |||
| GenBank mRNA | AK151613.1 | - | 3 | 661 | ||
| BC023677.1 | - | 3 | 648 | 5354497 | ||
| BC023774.1 | - | 3 | 670 | |||
| BQ713545.1 | - | 2 | 42 | |||
| BU053928.1 | - | 3 | 673 | |||
| CV563343.1 | - | 2 | 244 | |||