Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145363.2 | NP_663338.2 | 2 | 367 | ||
GenBank mRNA | AK084214.1 | - | 2 | 314 | ||
AK148037.1 | - | 2 | 310 | |||
AK148155.1 | - | 3 | 450 | |||
AK148383.1 | - | 2 | 303 | |||
AK182989.1 | - | 1 | 133 | |||
AK194285.1 | - | 1 | 131 | |||
BC019991.1 | - | 2 | 305 | 4189407 | ||
CK625019.1 | - | 2 | 367 |