Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145402.3 | NP_663377.1 | 3 | 1041 | ||
GenBank mRNA | AK077758.1 | - | 3 | 1041 | ||
AK164026.1 | - | 3 | 983 | |||
BC003277.1 | - | 3 | 905 | 3582174 |