siRNA Details
Gene Symbol: Lpcat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145376.5 | NP_663351.3 | 4 | 559 | ||
GenBank mRNA | AB244717.2 | - | 4 | 559 | ||
AK046079.1 | - | 2 | 354 | |||
AK046507.1 | - | 2 | 354 | |||
AK079946.1 | - | 717 | ||||
AK081865.1 | - | 4 | 717 | |||
AK132379.1 | - | 4 | 693 | |||
AK155286.1 | - | 4 | 723 | |||
AK169190.1 | - | 4 | 692 | |||
AK170723.1 | - | 4 | 712 | |||
AK171582.1 | - | 4 | 712 | |||
BC005662.1 | - | 2 | 89 | |||
BC066809.1 | - | 4 | 508 | 30475544 |

More Information
Panther Classification:
Molecular Function -






