Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145488.1 | NP_663463.1 | 1, 2 | 932 | ||
GenBank mRNA | AK030565.1 | - | 1 | 2438 | ||
AK083898.1 | - | 1, 2 | 920 | |||
AK143173.1 | - | 1, 2 | 920 | |||
AK220525.1 | - | 1, 2 | 913 | |||
BC003424.1 | - | 1, 2 | 932 | 2609884 |