Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145478.2 | NP_663453.1 | 2 | 581 | ||
GenBank mRNA | AK079623.1 | - | 2 | 301 | ||
AK149478.1 | - | 2 | 301 | |||
AK159399.1 | - | 2 | 300 | |||
AK159600.1 | - | 2 | 300 | |||
AK159670.1 | - | 2 | 277 | |||
AK168014.1 | - | 2 | 590 | |||
AY026239.1 | - | 2 | 329 | |||
BC017621.1 | - | 2 | 581 | 4924687 | ||
BC026639.1 | - | 2 | 270 | 4985551 |