Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025431.1 | NP_001020602.1 | 4 | 622 | ||
NM_145534.2 | NP_663509.2 | 3 | 924 | |||
GenBank mRNA | AB093275.1 | - | 3 | 924 | ||
AK052530.1 | - | 4 | 583 | |||
AK161924.1 | - | 4 | 579 | |||
BC018174.1 | - | 3 | 901 | 4011475 | ||
BC062968.1 | - | 3 | 620 | 6408511 |