Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145711.4 | NP_663757.3 | 3 | 504 | ||
GenBank mRNA | AF472514.1 | - | 3 | 300 | ||
AK029694.1 | - | 3 | 301 | |||
AK038671.1 | - | 3 | 507 | |||
AK051947.1 | - | 4 | 482 | |||
AK133397.1 | - | 3 | 301 | |||
AK161578.1 | - | 3 | 289 | |||
AK161718.1 | - | 3 | 264 | |||
BC080732.1 | - | 3 | 421 | 6518356 |