Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145614.4 | NP_663589.3 | 14 | 2310 | ||
GenBank mRNA | AK032124.1 | - | 14 | 2311 | ||
AK043771.1 | - | 14 | 1999 | |||
AK166090.1 | - | 14 | 2081 | |||
BC003202.1 | - | 3 | 438 |