siRNA Details
Gene Symbol: Mgat4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145926.3 | NP_666038.3 | 9, 10 | 1351 | ||
GenBank mRNA | AB053218.1 | - | 9, 10 | 1053 | ||
AK155336.1 | - | 9, 10 | 1352 | |||
AK192721.1 | - | 1, 2 | 74 | |||
BC026638.1 | - | 4, 5 | 459 | 4984907 | ||
BC031613.1 | - | 3, 4 | 329 | 4216720 | ||
CX205743.1 | - | 6, 7 | 611 | |||
CX206051.1 | - | 1, 2 | 110 |
