Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Pank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145962.2 | NP_666074.1 | 2 | 553 | ||
GenBank mRNA | AK039865.1 | - | 2 | 555 | ||
AK136305.1 | - | 2 | 243 | |||
BC027089.1 | - | 2 | 165 | 5355443 | ||
BC032188.1 | - | 2 | 599 |