siRNA Details
Gene Symbol: Mgat4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145926.3 | NP_666038.3 | 14 | 1840 | ||
GenBank mRNA | AB053218.1 | - | 14 | 1542 | ||
BC026638.1 | - | 9 | 948 | 4984907 | ||
BC031613.1 | - | 8 | 818 | 4216720 | ||
CX206051.1 | - | 6 | 599 | |||
W66623.1 | - | 3 | 170 |
