Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177374.1 | NP_001170845.1 | 18 | 2381 | ||
NM_146078.3 | NP_666190.2 | 18 | 2381 | |||
GenBank mRNA | AK083320.1 | - | 18 | 2520 | ||
AK122254.2 | - | 17 | 3263 | |||
AK143202.1 | - | 18 | 2381 | |||
AK147445.1 | - | 18 | 2370 | |||
AY061885.1 | - | 18 | 2068 | |||
AY280958.1 | - | 18 | 2096 | |||
BC031403.1 | - | 3 | 175 | 4935220 | ||
BC044903.1 | - | 4 | 316 | |||
BC075642.1 | - | 19 | 2195 | 30544183 |