siRNA Details
Gene Symbol: Gabrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146017.3 | NP_666129.1 | 10 | 2262 | ||
GenBank mRNA | AK028946.1 | - | 10 | 2262 | ||
BC023693.1 | - | 10 | 2249 | 5344479 | ||
BC027245.1 | - | 10 | 2225 | 3602400 | ||
BC031196.1 | - | 10 | 2236 | 5059564 |

More Information
Panther Classification:
Molecular Function -






