Search
Search

Fads1| Gene Symbol: Fads1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146094.2 | NP_666206.1 | 2 | 415 | ||
| GenBank mRNA | AB072976.1 | - | 2 | 330 | ||
| AF061504.1 | - | 2 | 317 | |||
| AK080706.1 | - | 2 | 319 | |||
| AK083959.1 | - | 2 | 277 | |||
| AK154367.1 | - | 2 | 365 | |||
| BC022139.1 | - | 2 | 360 | 5100199 | ||
| BC026831.1 | - | 2 | 382 | 4239695 | ||
| BC026848.1 | - | 2 | 355 | 4196030 | ||
| BC063053.1 | - | 2 | 275 | 5696722 | ||
| KF987079.1 | - | 2 | 299 | |||