Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164573.1 | NP_001158045.1 | 25 | 2510 | ||
GenBank mRNA | AF426467.1 | - | 3 | 191 | ||
AK014505.1 | - | 4 | 440 | |||
BC144867.1 | - | 4 | 415 | 9053415 | ||
BC144868.1 | - | 4 | 415 | |||
BC144869.1 | - | 4 | 415 | |||
BC144870.1 | - | 4 | 391 | 9053418 | ||
BC144872.1 | - | 4 | 415 |