Search
Search

Pbxip1| Gene Symbol: Pbxip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146131.2 | NP_666243.1 | 3 | 257 | ||
| GenBank mRNA | AK028754.1 | - | 3 | 218 | ||
| AK028858.1 | - | 3 | 255 | |||
| AK159422.1 | - | 3 | 244 | |||
| AK159448.1 | - | 3 | 244 | |||
| AK159633.1 | - | 3 | 234 | |||
| AK160103.1 | - | 3 | 238 | |||
| AK170318.1 | - | 3 | 213 | |||
| BC026838.1 | - | 2 | 192 | 4460993 | ||
| BC027771.1 | - | 3 | 227 | 5344900 | ||
| BC034200.1 | - | 3 | 198 | 4986595 | ||