Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tesk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146151.4 | NP_666263.3 | 2, 3 | 613 | ||
GenBank mRNA | AK031814.1 | - | 2, 3 | 606 | ||
AK047755.1 | - | 2, 3 | 614 | |||
AK053368.1 | - | 682 | ||||
BC019149.1 | - | 1, 2 | 346 | 5053345 | ||
BC029766.1 | - | 2, 3 | 591 | 4947564 |