Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PNPLA7 SIRNA Assays ›
          Gene Symbol:
          Pnpla7
          Gene Name:
          patatin-like phospholipase domain containing 7
          Gene Aliases:
          BC027342, E430013P11Rik, Nre
          Chromosome Location:
          Chr.2: 24975171 - 25054072 on Build GRCm38
          UniGene ID:
          Mm.389243
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 91084

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pnpla7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_146251.4 NP_666363.3 5 591
          GenBank mRNA AK038505.1 - 5 477
          AK088362.1 - 5 537
          AK148380.1 - 5 565
          AK170417.1 - 5 537
          BB618610.1 - 5 566
          BC021312.1 - 5 447
          BC141365.1 - 5 375 9055978
          BC141366.1 - 5 375 9055979
          EU293208.1 - 4 262
          EU368675.1 - 4 262
          EU402948.2 - 4 262

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          esterase

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          metabolic process
          developmental process
          phospholipase activity
          lysophospholipase activity
          prenylcysteine methylesterase activity
          lipase activity
          hydrolase activity
          1-oxa-2-oxocycloheptane lactonase activity
          sulfolactone hydrolase activity
          3,4-dihydrocoumarin hydrolase activity
          butyrolactone hydrolase activity
          endosulfan lactone lactonase activity
          L-ascorbate 6-phosphate lactonase activity
          Ser-tRNA(Thr) hydrolase activity
          Ala-tRNA(Pro) hydrolase activity
          Cys-tRNA(Pro) hydrolase activity
          Ser(Gly)-tRNA(Ala) hydrolase activity
          all-trans-retinyl-palmitate hydrolase, all-trans-retinol forming activity
          retinyl-palmitate esterase activity
          carboxylic ester hydrolase activity
          mannosyl-oligosaccharide 1,6-alpha-mannosidase activity
          mannosyl-oligosaccharide 1,3-alpha-mannosidase activity
          methyl indole-3-acetate esterase activity
          methyl salicylate esterase activity
          methyl jasmonate esterase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0