Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146748.1 | NP_666959.1 | 1 | 363 | ||
GenBank mRNA | BC128005.1 | - | 1 | 367 | 40135960 |