siRNA Details
Gene Symbol: Olfr1214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146897.2 | NP_667108.2 | 1 | 271 |

Gene Symbol: Olfr1214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146897.2 | NP_667108.2 | 1 | 271 |