Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr295 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146851.2 | NP_667062.2 | 1 | 193 |
Gene Symbol: Olfr292 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146620.2 | NP_666831.2 | 2 | 343 | ||
GenBank mRNA | CB173961.1 | - | 2 | 343 |