Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012.1 | NP_001003.1 | 3 | 182 | ||
GenBank mRNA | AB062401.1 | - | 2 | 161 | ||
AF116700.1 | - | 2 | 77 | |||
AK311854.1 | - | 3 | 182 | |||
BC070875.1 | - | 2 | 169 | 6527505 |