Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146955.1 | NP_667166.1 | 1 | 828 | ||
GenBank mRNA | AF102539.1 | - | 1 | 636 | ||
BC106784.1 | - | 1 | 828 | 40041376 | ||
BC106785.1 | - | 1 | 826 | 40041377 |