Search
Search

TUBG1| Gene Symbol: TUBG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001070.4 | NP_001061.2 | 3, 4 | 736 | ||
| GenBank mRNA | AK313339.1 | - | 3, 4 | 405 | ||
| BC000619.2 | - | 3, 4 | 400 | 3345973 | ||
| BT019931.1 | - | 3, 4 | 338 | |||
| CR407642.1 | - | 3, 4 | 338 | |||
| M61764.1 | - | 3, 4 | 362 | |||