Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB049454.2 | - | 4 | 359 | ||
AB100426.1 | - | 4 | 359 | |||
BC120809.1 | - | 3 | 280 | 40129732 |