Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npas4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153553.4 | NP_705781.1 | 1 | 319 | ||
GenBank mRNA | AB049835.1 | - | 1 | 214 | ||
AK032607.1 | - | 1 | 319 | |||
AY730724.1 | - | 1 | 346 | |||
BC059815.1 | - | 1 | 664 | |||
BC079544.1 | - | 1 | 664 | |||
BC129861.1 | - | 1 | 255 | 40110003 |