Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aaas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153416.2 | NP_700465.2 | 12 | 1248 | ||
GenBank mRNA | AK034591.1 | - | 12 | 1143 | ||
AK079849.1 | - | 12 | 1185 | |||
AK083401.1 | - | 12 | 1162 | |||
AK083537.1 | - | 12 | 1169 | |||
AK087134.1 | - | 12 | 1224 | |||
AK088247.1 | - | 12 | 1165 | |||
AK134406.1 | - | 12 | 1139 | |||
AK165243.1 | - | 12 | 1163 | |||
AK166118.1 | - | 12 | 1163 | |||
AK169523.1 | - | 12 | 1200 | |||
BC018191.1 | - | 12 | 1149 | 3994248 | ||
BC025501.1 | - | 12 | 1195 | 5321013 | ||
BC026604.1 | - | 11 | 1702 |