Search
Search

Insig1| Gene Symbol: Insig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153526.5 | NP_705746.1 | 5 | 2172 | ||
| GenBank mRNA | AK007471.1 | - | 1 | 515 | ||
| AK088380.1 | - | 5 | 2212 | |||
| AK135941.1 | - | 5 | 2222 | |||
| AK135989.1 | - | 5 | 2225 | |||
| AK139913.1 | - | 5 | 2221 | |||
| AK151186.1 | - | 5 | 2222 | |||
| BC079559.1 | - | 5 | 2172 | |||
| BC132167.1 | - | 5 | 2173 | 40130444 | ||
| BC132499.1 | - | 5 | 2175 | 40130776 | ||