Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172064.1 | NP_001165535.1 | 4 | 770 | ||
NM_001172065.1 | NP_001165536.1 | 4 | 770 | |||
NM_153777.2 | NP_722472.1 | 4 | 356 | |||
GenBank mRNA | AK017603.1 | - | 4 | 744 | ||
AK035366.1 | - | 2 | 3116 | |||
AK220237.1 | - | 4 | 724 | |||
BC027807.1 | - | 4 | 366 | 5324022 |