Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170756.2 | NP_739562.1 | 3 | 611 | ||
GenBank mRNA | AK051741.1 | - | 3 | 557 | ||
AK122364.1 | - | 1 | 1253 | |||
AK144826.1 | - | 3 | 583 | |||
AK156188.1 | - | 3 | 612 | |||
AK162743.1 | - | 3 | 611 | |||
AK170356.1 | - | 3 | 612 | |||
AK171240.1 | - | 3 | 585 | |||
AK171664.1 | - | 3 | 589 | |||
AK171852.1 | - | 3 | 601 | |||
AK191359.1 | - | 1 | 22 | |||
BC034597.1 | - | 3 | 600 | 5363513 |