Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sulf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198565.1 | NP_001185494.1 | 4 | 584 | ||
NM_001198566.1 | NP_001185495.1 | 3 | 489 | |||
NM_172294.1 | NP_758498.1 | 4 | 568 | |||
GenBank mRNA | AK028285.1 | - | 3 | 388 | ||
AK037389.1 | - | 4 | 610 | |||
AK045002.1 | - | 4 | 483 | |||
AK054518.1 | - | 3 | 804 | |||
AK078756.1 | - | 4 | 573 | |||
AK081929.1 | - | 3 | 391 | |||
AK129278.1 | - | 3 | 390 | |||
AY101178.1 | - | 4 | 593 | |||
BC034547.1 | - | 4 | 568 | 4500954 | ||
BC049276.1 | - | 4 | 568 | 4500954 | ||
CD348948.1 | - | 4 | 584 |